5-Chloroisophthalonitrile

Lieferant: Apollo Scientific

OR470922-1G
APOSOR470922-1GEA 1120 EUR
APOSOR470922-1G
5-Chloroisophthalonitrile
5-Chloroisophthalonitrile
Formel: C₈H₃ClN₂
Lagertemperatur: Raumtemperatur
MDL: MFCD27959539
CAS-Nummer: 109292-88-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR