4,6-Dimethylpyridine-3-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR470924-1G
APOSOR470924-1GEA 1290 EUR
APOSOR470924-1G
4,6-Dimethylpyridine-3-boronic acid pinacol ester
4,6-Dimethylpyridine-3-boronic acid pinacol ester
Formel: C₁₃H₂₀BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18719934
CAS-Nummer: 1622217-32-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR