2-Ethylpyridine-3-boronic acid pinacol ester

Lieferant: Apollo Scientific

OR470926-1G
APOSOR470926-1GEA 1540 EUR
APOSOR470926-1G
2-Ethylpyridine-3-boronic acid pinacol ester
2-Ethylpyridine-3-boronic acid pinacol ester
Formel: C₁₃H₂₀BNO₂
Molecular Weight: 233,114 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13182289
CAS-Nummer: 1494670-02-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR