3-(3-Methoxybenzyl)pyrrolidine hydrochloride

Lieferant: Apollo Scientific

OR470928-1G
APOSOR470928-1GEA 804 EUR
APOSOR470928-1G
3-(3-Methoxybenzyl)pyrrolidine hydrochloride
3-(3-Methoxybenzyl)pyrrolidine hydrochloride
Formel: C₁₂H₁₇NO·ClH
Molecular Weight: 227,73 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986825
CAS-Nummer: 2006277-93-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR