(R)-3-(Cbz-amino)-2-piperidone

Lieferant: Apollo Scientific

OR470929-1G
APOSOR470929-1GEA 561 EUR
APOSOR470929-1G
(R)-3-(Cbz-amino)-2-piperidone
(R)-3-(Cbz-amino)-2-piperidone
Formel: C₁₃H₁₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09264105
CAS-Nummer: 722499-65-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR