Ethyl 3-(5-Chloro-2-nitrophenyl)propanoate

Lieferant: Apollo Scientific

OR470930-1G
APOSOR470930-1GEA 1150 EUR
APOSOR470930-1G
Ethyl 3-(5-Chloro-2-nitrophenyl)propanoate
Ethyl 3-(5-Chloro-2-nitrophenyl)propanoate
Formel: C₁₁H₁₂ClNO₄
Molecular Weight: 257,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986823
CAS-Nummer: 1803762-10-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR