Ethyl 3-(3-Chloro-2-nitrophenyl)propanoate

Lieferant: Apollo Scientific

OR470932-1G
APOSOR470932-1GEA 1150 EUR
APOSOR470932-1G
Ethyl 3-(3-Chloro-2-nitrophenyl)propanoate
Ethyl 3-(3-Chloro-2-nitrophenyl)propanoate
Formel: C₁₁H₁₂ClNO₄
Molecular Weight: 257,67 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986821
CAS-Nummer: 1803762-08-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR