Ethyl 5-Bromoisoquinoline-1-carboxylate
Lieferant: Apollo Scientific
OR470933-1G
APOSOR470933-1GEA
1150
EUR
APOSOR470933-1G
Ethyl 5-Bromoisoquinoline-1-carboxylate
Ethyl 5-Bromoisoquinoline-1-carboxylate
Formel:
C₁₂H₁₀BrNO₂ Molecular Weight: 280,117 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD27986819 CAS-Nummer: 1111311-65-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...