1-Bromo-1-nitrocyclobutane

Lieferant: Apollo Scientific

OR470936-1G
APOSOR470936-1GEA 499 EUR
APOSOR470936-1G
1-Bromo-1-nitrocyclobutane
1-Bromo-1-nitrocyclobutane
Formel: C₄H₆BrNO₂
Molecular Weight: 180 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD27986827
CAS-Nummer: 51175-81-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR