4-Hydroxy-2,5-dimethylbenzoic acid

Lieferant: Apollo Scientific

OR470938-1G
APOSOR470938-1GEA 1010 EUR
APOSOR470938-1G
4-Hydroxy-2,5-dimethylbenzoic acid
4-Hydroxy-2,5-dimethylbenzoic acid
Formel: C₉H₁₀O₃
Molecular Weight: 166,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09999327
CAS-Nummer: 27021-04-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR