4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester

Lieferant: Apollo Scientific

OR470939-1G
APOSOR470939-1GEA 1060 EUR
APOSOR470939-1G
4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester
4-(Hydroxymethyl)-3-methylphenylboronic acid pinacol ester
Formel: C₁₄H₂₁BO₃
Molecular Weight: 248,126 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18732622
CAS-Nummer: 1160430-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR