3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
(2-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol
OR470940-1G
APOSOR470940-1GEA
1060
EUR
APOSOR470940-1G
3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester
3-Chloro-4-(hydroxymethyl)phenylboronic acid pinacol ester
Formel:
C₁₃H₁₈BClO₃ Molecular Weight: 268.544 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18732618 CAS-Nummer: 1051316-34-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...