2,6-Dimethylpyridine-3-boronic acid pinacol ester
Lieferant: Apollo Scientific
Synonyme:
2,6-Dimethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
OR470942-1G
APOSOR470942-1GEA
1090
EUR
APOSOR470942-1G
2,6-Dimethylpyridine-3-boronic acid pinacol ester
2,6-Dimethylpyridine-3-boronic acid pinacol ester
Formel:
C₁₃H₂₀BNO₂ Molecular Weight: 233,114 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13195066 CAS-Nummer: 693774-10-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...