1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid

Lieferant: Apollo Scientific

OR470943-1G OR470943-250MG
APOSOR470943-1GEA 804 EUR
APOSOR470943-1G APOSOR470943-250MG
1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid
1-Oxo-1,2-dihydroisoquinoline-3-carboxylic acid
Formel: C₁₀H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD01845162
CAS-Nummer: 7509-13-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR