3-(3-Bromopropoxy)benzaldehyde

Lieferant: Apollo Scientific

OR470946-1G
APOSOR470946-1GEA 601 EUR
APOSOR470946-1G
3-(3-Bromopropoxy)benzaldehyde
3-(3-Bromopropoxy)benzaldehyde
Formel: C₁₀H₁₁BrO₂
Molecular Weight: 243,097 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06656204
CAS-Nummer: 144707-69-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR