6-Amino-9-(tetrahydropyran-2-yl)-9H-purine

Lieferant: Apollo Scientific

Synonyme: 9-(Tetrahydro-2H-pyran-2-yl)-9H-purin-6-amine

OR470947-1G OR470947-250MG OR470947-5G
APOSOR470947-1GEA 140 EUR
APOSOR470947-1G APOSOR470947-250MG APOSOR470947-5G
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine
6-Amino-9-(tetrahydropyran-2-yl)-9H-purine
Formel: C₁₀H₁₃N₅O
Lagertemperatur: Raumtemperatur
MDL: MFCD00213333
CAS-Nummer: 7306-67-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR