2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one

Lieferant: Apollo Scientific

OR470949-1G
APOSOR470949-1GEA 783 EUR
APOSOR470949-1G
2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one
2-(Methylthio)-5,6-dihydropyrimidin-4(1H)-one
Formel: C₅H₈N₂OS
Molecular Weight: 144,195 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19224225
CAS-Nummer: 100185-53-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR