2-Hydroxy-6-methylpyridine-3-boronic acid

Lieferant: Apollo Scientific

OR470951-1G
APOSOR470951-1GEA 1540 EUR
APOSOR470951-1G
2-Hydroxy-6-methylpyridine-3-boronic acid
2-Hydroxy-6-methylpyridine-3-boronic acid
Formel: C₆H₈BNO₃
Molecular Weight: 152,944 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16611421
CAS-Nummer: 1279715-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR