(Ethylsulphonyl)acetic acid
Lieferant: Apollo Scientific
OR470954-1G
OR470954-5G
APOSOR470954-1GEA
306
EUR
APOSOR470954-1G
APOSOR470954-5G
(Ethylsulphonyl)acetic acid
(Ethylsulphonyl)acetic acid
Formel:
C₄H₈O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD09864550 CAS-Nummer: 141811-44-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...