N-[(4-Pyridyl)methyl]cyclopropanamine

Lieferant: Apollo Scientific

OR470955-5G
APOSOR470955-5GEA 529 EUR
APOSOR470955-5G
N-[(4-Pyridyl)methyl]cyclopropanamine
N-[(4-Pyridyl)methyl]cyclopropanamine
Formel: C₉H₁₂N₂
Molecular Weight: 148,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03821870
CAS-Nummer: 193153-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR