N-[(3-Pyridyl)methyl]-2-propanamine

Lieferant: Apollo Scientific

Synonyme: N-(Pyridin-3-ylmethyl)propan-2-amine

OR470956-5G
APOSOR470956-5GEA 529 EUR
APOSOR470956-5G
N-[(3-Pyridyl)methyl]-2-propanamine
N-[(3-Pyridyl)methyl]-2-propanamine
Formel: C₉H₁₄N₂
Molecular Weight: 150,221 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00716868
CAS-Nummer: 19730-12-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR