3-Hydroxy-5-(methylsulphonamido)benzoic acid
Lieferant: Apollo Scientific
OR470958-1G
APOSOR470958-1GEA
499
EUR
APOSOR470958-1G
3-Hydroxy-5-(methylsulphonamido)benzoic acid
3-Hydroxy-5-(methylsulphonamido)benzoic acid
Formel:
C₈H₉NO₅S Molecular Weight: 231,226 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD16999874 CAS-Nummer: 1243364-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...