3-Acetamido-5-hydroxybenzoic acid

Lieferant: Apollo Scientific

Synonyme: 3-Acetamido-5-hydroxybenzoic Acid

OR470959-1G
APOSOR470959-1GEA 284 EUR
APOSOR470959-1G
3-Acetamido-5-hydroxybenzoic acid
3-Acetamido-5-hydroxybenzoic acid
Formel: C₉H₉NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD20654260
CAS-Nummer: 93561-88-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR