3-Hydroxy-5-(methylamino)benzoic acid

Lieferant: Apollo Scientific

OR470960-1G
APOSOR470960-1GEA 357 EUR
APOSOR470960-1G
3-Hydroxy-5-(methylamino)benzoic acid
3-Hydroxy-5-(methylamino)benzoic acid
Formel: C₈H₉NO₃
Molecular Weight: 167,162 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD16998481
CAS-Nummer: 89611-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR