2-[(1-Imidazolyl)methyl]aniline

Lieferant: Apollo Scientific

OR470962-5G
APOSOR470962-5GEA 601 EUR
APOSOR470962-5G
2-[(1-Imidazolyl)methyl]aniline
2-[(1-Imidazolyl)methyl]aniline
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09814189
CAS-Nummer: 61292-50-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR