2-[(1-Pyrazolyl)methyl]aniline

Lieferant: Apollo Scientific

OR470963-5G
APOSOR470963-5GEA 601 EUR
APOSOR470963-5G
2-[(1-Pyrazolyl)methyl]aniline
2-[(1-Pyrazolyl)methyl]aniline
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,214 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08572198
CAS-Nummer: 956533-57-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR