3-Pyrazol-1-ylmethylphenylamine

Lieferant: Apollo Scientific

OR470964-5G
APOSOR470964-5GEA 713 EUR
APOSOR470964-5G
3-Pyrazol-1-ylmethylphenylamine
3-Pyrazol-1-ylmethylphenylamine
Formel: C₁₀H₁₁N₃
Molecular Weight: 173.22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422525
CAS-Nummer: 892502-09-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR