4-(1H-Pyrazol-1-ylmethyl)aniline

Lieferant: Apollo Scientific

Synonyme: 4-((1H-Pyrazol-1-yl)methyl)aniline

OR470965-5G
APOSOR470965-5GEA 342 EUR
APOSOR470965-5G
4-(1H-Pyrazol-1-ylmethyl)aniline
4-(1H-Pyrazol-1-ylmethyl)aniline
Formel: C₁₀H₁₁N₃
Molecular Weight: 173,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03422514
CAS-Nummer: 142335-61-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR