Ethyl 6-(Isobutylamino)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

OR470966-1G
APOSOR470966-1GEA 499 EUR
APOSOR470966-1G
Ethyl 6-(Isobutylamino)pyridazine-3-carboxylate
Ethyl 6-(Isobutylamino)pyridazine-3-carboxylate
Formel: C₁₁H₁₇N₃O₂
Molecular Weight: 223,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014560
CAS-Nummer: 1823784-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR