Methyl 6-(Isobutylamino)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

OR470967-1G
APOSOR470967-1GEA 499 EUR
APOSOR470967-1G
Methyl 6-(Isobutylamino)pyridazine-3-carboxylate
Methyl 6-(Isobutylamino)pyridazine-3-carboxylate
Formel: C₁₀H₁₅N₃O₂
Molecular Weight: 209.245 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12152728
CAS-Nummer: 1184230-38-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR