6-(Isobutylamino)pyridazine-3-carboxylic acid

Lieferant: Apollo Scientific

OR470968-1G
APOSOR470968-1GEA 713 EUR
APOSOR470968-1G
6-(Isobutylamino)pyridazine-3-carboxylic acid
6-(Isobutylamino)pyridazine-3-carboxylic acid
Formel: C₉H₁₃N₃O₂
Molecular Weight: 195,218 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12152628
CAS-Nummer: 1178687-41-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR