Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Lieferant: Apollo Scientific
OR470969-1G
APOSOR470969-1GEA
499
EUR
APOSOR470969-1G
Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Ethyl 6-(Isopropylamino)pyridazine-3-carboxylate
Formel:
C₁₀H₁₅N₃O₂ Molecular Weight: 209,245 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD28014559 CAS-Nummer: 1823359-03-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...