6-(Isopropylamino)pyridazine-3-carboxylic acid

Lieferant: Apollo Scientific

OR470971-1G
APOSOR470971-1GEA 713 EUR
APOSOR470971-1G
6-(Isopropylamino)pyridazine-3-carboxylic acid
6-(Isopropylamino)pyridazine-3-carboxylic acid
Formel: C₈H₁₁N₃O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD12152624
CAS-Nummer: 1183803-85-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR