Ethyl 6-(Cyclopropylamino)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

OR470972-1G
APOSOR470972-1GEA 713 EUR
APOSOR470972-1G
Ethyl 6-(Cyclopropylamino)pyridazine-3-carboxylate
Ethyl 6-(Cyclopropylamino)pyridazine-3-carboxylate
Formel: C₁₀H₁₃N₃O₂
Molecular Weight: 207,229 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014558
CAS-Nummer: 1823333-44-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR