Methyl 6-(cyclopropylamino)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

OR470973-1G
APOSOR470973-1GEA 713 EUR
APOSOR470973-1G
Methyl 6-(cyclopropylamino)pyridazine-3-carboxylate
Methyl 6-(cyclopropylamino)pyridazine-3-carboxylate
Formel: C₉H₁₁N₃O₂
Molecular Weight: 193,203 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12152727
CAS-Nummer: 1183253-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR