6-(Cyclopropylamino)pyridazine-3-carboxylic acid

Lieferant: Apollo Scientific

OR470974-1G
APOSOR470974-1GEA 713 EUR
APOSOR470974-1G
6-(Cyclopropylamino)pyridazine-3-carboxylic acid
6-(Cyclopropylamino)pyridazine-3-carboxylic acid
Formel: C₈H₉N₃O₂
Molecular Weight: 179,176 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12152627
CAS-Nummer: 1178768-37-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR