Ethyl 6-(Ethylamino)pyridazine-3-carboxylate
Lieferant: Apollo Scientific
OR470975-1G
APOSOR470975-1GEA
499
EUR
APOSOR470975-1G
Ethyl 6-(Ethylamino)pyridazine-3-carboxylate
Ethyl 6-(Ethylamino)pyridazine-3-carboxylate
Formel:
C₉H₁₃N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28014557 CAS-Nummer: 1823915-03-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...