Methyl 6-(ethylamino)pyridazine-3-carboxylate
Lieferant: Apollo Scientific
OR470976-1G
APOSOR470976-1GEA
499
EUR
APOSOR470976-1G
Methyl 6-(ethylamino)pyridazine-3-carboxylate
Methyl 6-(ethylamino)pyridazine-3-carboxylate
Formel:
C₈H₁₁N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD12149171 CAS-Nummer: 1179655-77-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...