6-(Ethylamino)pyridazine-3-carboxylic acid
Lieferant: Apollo Scientific
OR470977-1G
APOSOR470977-1GEA
713
EUR
APOSOR470977-1G
6-(Ethylamino)pyridazine-3-carboxylic acid
6-(Ethylamino)pyridazine-3-carboxylic acid
Formel:
C₇H₉N₃O₂ Molecular Weight: 167,165 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12152631 CAS-Nummer: 1183245-80-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...