Ethyl 6-(Methylamino)pyridazine-3-carboxylate

Lieferant: Apollo Scientific

OR470978-1G
APOSOR470978-1GEA 499 EUR
APOSOR470978-1G
Ethyl 6-(Methylamino)pyridazine-3-carboxylate
Ethyl 6-(Methylamino)pyridazine-3-carboxylate
Formel: C₈H₁₁N₃O₂
Molecular Weight: 181,192 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014556
CAS-Nummer: 365413-16-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR