Methyl 3-Bromo-5-chlorobenzimidate hydrochloride

Lieferant: Apollo Scientific

OR470979-1G
APOSOR470979-1GEA 357 EUR
APOSOR470979-1G
Methyl 3-Bromo-5-chlorobenzimidate hydrochloride
Methyl 3-Bromo-5-chlorobenzimidate hydrochloride
Formel: C₈H₇BrClNO·ClH
Molecular Weight: 284,965 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014555
CAS-Nummer: 1823380-00-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR