Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride

Lieferant: Apollo Scientific

OR470981-1G
APOSOR470981-1GEA 306 EUR
APOSOR470981-1G
Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride
Methyl 3-[Imino(methoxy)methyl]benzoate hydrochloride
Formel: C₁₀H₁₁NO₃·ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD10697976
CAS-Nummer: 1154063-16-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR