Methyl 2-[Imino(methoxy)methyl]benzoate hydrochloride

Lieferant: Apollo Scientific

OR470982-1G
APOSOR470982-1GEA 397 EUR
APOSOR470982-1G
Methyl 2-[Imino(methoxy)methyl]benzoate hydrochloride
Methyl 2-[Imino(methoxy)methyl]benzoate hydrochloride
Formel: C₁₀H₁₁NO₃·ClH
Molecular Weight: 229,66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014553
CAS-Nummer: 1823954-95-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR