Methyl 2-Bromobenzimidate hydrochloride

Lieferant: Apollo Scientific

OR470984-1G
APOSOR470984-1GEA 306 EUR
APOSOR470984-1G
Methyl 2-Bromobenzimidate hydrochloride
Methyl 2-Bromobenzimidate hydrochloride
Formel: C₈H₈BrNO·ClH
Molecular Weight: 250,52 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28014552
CAS-Nummer: 2006277-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR