4-[(Cyclopropylamino)sulphonyl]benzoic acid
Lieferant: Apollo Scientific
OR470987-5G
APOSOR470987-5GEA
713
EUR
APOSOR470987-5G
4-[(Cyclopropylamino)sulphonyl]benzoic acid
4-[(Cyclopropylamino)sulphonyl]benzoic acid
Formel:
C₁₀H₁₁NO₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD04035198 CAS-Nummer: 436092-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...