4-[(Ethylamino)sulphonyl]benzoic acid

Lieferant: Apollo Scientific

OR470990-1G
APOSOR470990-1GEA 397 EUR
APOSOR470990-1G
4-[(Ethylamino)sulphonyl]benzoic acid
4-[(Ethylamino)sulphonyl]benzoic acid
Formel: C₉H₁₁NO₄S
Molecular Weight: 229.253 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06357875
CAS-Nummer: 10252-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR