2-Chloro-4-ethoxybenzoic acid

Lieferant: Apollo Scientific

OR470996-5G
APOSOR470996-5GEA 601 EUR
APOSOR470996-5G
2-Chloro-4-ethoxybenzoic acid
2-Chloro-4-ethoxybenzoic acid
Formel: C₉H₉ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD17256595
CAS-Nummer: 334018-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR