3-Chloro-4-ethoxybenzoic acid ≥98%

Lieferant: Apollo Scientific

OR470997-250MG
APOSOR470997-250MGEA 75.4 EUR
APOSOR470997-250MG
3-Chloro-4-ethoxybenzoic acid ≥98%
3-Chloro-4-ethoxybenzoic acid
Formel: C₉H₉ClO₃
Molecular Weight: 200,619 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD02063066
CAS-Nummer: 213598-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR