Methyl 3-Amino-4,4-dimethylpentanoate

Lieferant: Apollo Scientific

OR470998-1G
APOSOR470998-1GEA 855 EUR
APOSOR470998-1G
Methyl 3-Amino-4,4-dimethylpentanoate
Methyl 3-Amino-4,4-dimethylpentanoate
Formel: C₈H₁₇NO₂
Molecular Weight: 159,226 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22532139
CAS-Nummer: 1273387-45-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR