2-Chloro-5-(2-thienyl)pyrazine

Lieferant: Apollo Scientific

OR471000-1G
APOSOR471000-1GEA 916 EUR
APOSOR471000-1G
2-Chloro-5-(2-thienyl)pyrazine
2-Chloro-5-(2-thienyl)pyrazine
Formel: C₈H₅ClN₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD08277628
CAS-Nummer: 912762-30-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR